Somso® Beauchene Skull
Supplier: MARCUS SOMMER
QS 9
470030-010EA 1982.12 USD
470030-010
Somso® Beauchene Skull
Models Human Biology Models
A classic Beauchene view.
  • Removable Pieces for Individual Study
  • Five-Year Manufacturer's Warranty
  • Life-Size Skull
The life-size skull has been carefully disarticulated and mounted in an “exploded” position on a clear plastic support to retain the bones’ spatial relationships.
Mounted on a stand with base, the model can be tilted back, so students can view the inside of the skull.