Somso® Peripheral Cross Section of a Stem Model
Supplier: MARCUS SOMMER
BOS 22/6
470029-850EA 722.51 USD
470029-850
Somso® Peripheral Cross Section of a Stem Model
Models Botany Models
See Microscopic Detail
  • 5 Year Manufacturers Warranty
This reproduction of a rambling Ranunculus is enlarged 450X. The model shows the epidermis with stomata, xylem, phloem, cambium, and more.