Somso® Fertilization of the Angiosperms
Supplier: MARCUS SOMMER
BOS 19
470029-844EA 635.59 USD
470029-844
Somso® Fertilization of the Angiosperms
Models Human Biology Models
Somso® premium quality models.
  • Five year warranty
Polygonum-type, enlarged 300 times, in SOMSO-Plast®. After Prof. Dr. W. Jung. Longitudinal section showing ovary with germinating pollen grains and embryo sac, stamen with pollen in sagittal section. Showing in detail the microscopic structure. On a green base with description key. In one piece.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.