Somso® Chloroplast Model
Supplier: MARCUS SOMMER
BOS 16/2
470029-842EA 1185.24 USD
470029-842
Somso® Chloroplast Model
Models Botany Models
Show Students Where Photosynthesis Occurs
  • Illustrates The Complex Internal Structure Of A Chloroplast
  • View Structures In Longitudinal Or Cross Section
  • 5 Year Manufacturers Warranty
Take apart the three-dimensional model of a chloroplast and remove the thylacoids and linking stroma to reveal the interior structure where photosynthesis occurs. Patterned after the latest electron microscopy interpretation, the structure is depicted in longitudinal and cross sections to show the interior of the chloroplast enlarged 60,000X; one quarter of the model is cut away to reveal the inner detail in three levels.