Somso® Fern Prothallium Model
Supplier: MARCUS SOMMER
BOS 14/5
470029-836EA 662.94 USD
470029-836
Somso® Fern Prothallium Model
Models Botany Models
Detailed Rendering Of Complete Fern Cycle
  • One Model Showing Aspects Of The Complete Life Cycle
  • 5 Year Manufacturers Warranty
Enlarged 45X, the Dryopteris fern prothallium features the antheridia and archegonia on the underside, and a developing fern emerging from a fertilized ovum on the top. It also shows the rhizoids that anchor the prothallium.