Rescue Critters® Critical Care Jerry
Supplier: THALES & CO.
110
470019-134EA 1835 USD
470019-134 470029-826
Rescue Critters® Critical Care Jerry
Models Zoology Models
A Complete Emergency Room Veterinary Training Manikin
  • Has working lungs and artificial pulse
  • Made to order
  • Included carrying case
Approximating a 60-70 pound dog, with a femoral manual pulse, Critical Care Jerry provides veterinary students the opportunity to practice numerous life-saving skills and complicated maneuvers, like aspirating air and fluid from the thoracic cavity. Includes IV Trainer Kit with forearm, IV pole and holder, two IV reservoir bags, and artificial training blood. It has the ability to simulate trauma and features jugular and vascular access. Realistic representations of the trachea, esophagus, and epiglottis are built into the airway.
These provide for the training of:
airway management
ventilation
endotracheal intubation
mouth-to-snout resuscitation
circulation with external chest compressions
Manikin comes with working lungs and artificial pulse and features for splinting and bandaging.
Included in the carrying case are a kneeling pad, endotracheal tube, syringe, brush, and 5 disposable lungs.
Contact WARD’S for a quote on other features available for custom installation in your Critical Care Jerry.
Ordering information: This item is made to order. Returns cannot be accepted.