Somso® Head of A Gnat
Supplier: MARCUS SOMMER
ZOS 48/3
470029-798EA 786.68 USD
470029-798
Somso® Head of A Gnat
Models Zoology Models
Somso® premium quality models.
  • 5 year warranty
Culex pipiens, head of a female gnat, modelled from nature. After Dr. E. Schicha. Enlarged approx. 80 times, in SOMSO-Plast® The long stylets, consisting of the labrum (upper lip), the paired mandibles, the paired maxillae, and the hypopharynx can be put together to demonstrate the function of the stylets. In one piece. On a stand with green base.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.