Somso® Liver Cell Model
Supplier: MARCUS SOMMER
JS 15
470029-708EA 1311.18 USD
470029-708
Somso® Liver Cell Model
Models Human Biology Models
Study Physiological Transport Within The Liver On A Cellular Level
  • 5 Year Manufacturers Warranty
Show organelles and the sophisticated transport systems for the flow of blood and bile with this transparent model. You could also use it to demonstrate the structure of a generalized animal cell.