Somso® Cochlea Section Model
Supplier: MARCUS SOMMER
DS 10
470029-658EA 781.95 USD
470029-658
Somso® Cochlea Section Model
Models Human Biology Models
A detailed view of the cochlea.
  • Detailed Model of a Complex Structure
  • Five-Year Manufacturer's Warranty
  • Model Displays the Mechanism of Sound Reception
The model shows the vestibular, cochlear, and tympanic ducts; Corti’s organ with hair cells and tectorial and basilar membranes; and spiral ganglia, nerves, and arteries. This model is a perfect complement to any general ear model because it renders one of the key structures of the ear, enlarged 350x, in great detail.
It also brings our cochlea microscope slide to three-dimensional life.