Somso® Comprehensive Functional Larynx Model
Supplier: MARCUS SOMMER
GS 10
470029-638EA 1058.6 USD
470029-638
Somso® Comprehensive Functional Larynx Model
Models Human Biology Models
Demonstrates Vocal Cord Movement
  • Vocal Cords Can Be Manipulated
  • Large Size For Easier Manipulation And Examination
  • 5 Year Manufacturers Warranty
Show how the larynx works with this 3X-enlarged, complete model featuring all structural components, including the hyoid bone, surrounding cartilage, and musculature. Students can re-create the motion of the larynx by increasing the tension on the drawstring to rotate the arytenoid cartilage, opening the vocal cords, making it perfect for lessons on speech, physiology, or anatomy.