Somso® Generalized Plant Cell Model
Supplier: MARCUS SOMMER
BOS 16
470029-606EA 253.85 USD
470029-606
Somso® Generalized Plant Cell Model
Models Molecular Biology Models
Excellent Introduction To Plants’ Microanatomy
  • General Plant Cell Structure In Somso Detail
  • Key Printed Directly On Base For Easy Reference
  • 5 Year Manufacturers Warranty
View the nucleus, chloroplasts, cell wall, and vacuoles of a plant cell, as well as discover the cell’s relationship to adjacent cells. The simplified model, enlarged 3,000X, is perfect for introducing students to the basic parts of any plant cell.