GPI Anatomicals® Canine 4-Stage Knee Model
Supplier: GPI ANATOMICALS
9051 9051
470029-478PK 165 USD
470029-478 470029-479
GPI Anatomicals® Canine 4-Stage Knee Model
Models Zoology Models
Demonstrate Osteoarthritis
Illustrate degenerative joint disease with this life-size set of four models. Models depict a disease-free (normal) knee highlighting important structures including ligaments, tendons and patella. The progressive degeneration of the articular surfaces and the development of osteophytes or bone spurs are demonstrated in the other three models.