GPI Anatomicals® Canine Skin Model
Supplier: GPI ANATOMICALS
9090
470029-468EA 145.8 USD
470029-468
GPI Anatomicals® Canine Skin Model
Models Zoology Models
Enlarged Cross-Section Of Canine Skin
This double-sided model reveals both healthy and infected layers of a common dog. One side features normal skin anatomy, extending from the epidermis down to the layer of subcutaneous far; while the opposite side demonstrates the progressing changes of a flea bite from wheal to infected pustule.