Ward's® Endocrine System Model
Supplier: The Science Source
810840
470029-450EA 492.05 USD
470029-450
Ward's® Endocrine System Model
Models Human Biology Models
A Ward's exclusive.
  • Cutaway views reveal the location and size of glands and structures in relation to other organs
  • Includes a key card
  • Custom molded using the highest quality nontoxic polymers and PVC
  • Hand-painted by skilled artists
This unique hand-painted model features cutaway views for observing the location and size of glands in relation to other organs. Endocrine structures include the thymus (appropriately sized for a child before involution), thyroid, pineal gland, pituitary gland, testis, umbilical and hepatic veins, and more.