Ward's® Bone Structure Model
Supplier: The Science Source
810660
470029-440EA 490.4 USD
470029-440
Ward's® Bone Structure Model
Models Human Biology Models
This model displays various layers simultaneously.
  • Simple Design Shows Detailed Bone Structure
  • Cutaway Section Shows Bone Structure in Multiple Angles
  • Designed by Wards
  • Enlarged 500x
  • Complete Haversian System Displayed
Enlarged 500x, the cross section displays the complete Haversian system with the lamellae cut away in steps to reveal the structural detail at each level.
The vasculature, nerve, lymph vessels, osteocytes, lacunae, and canaliculi are also on display.