Ward's® Blood Components Model Set
Supplier: The Science Source
810601
470029-436KT 1532.94 USD
470029-436
Ward's® Blood Components Model Set
Models Human Biology Models
Exceptional realism recreates details normally visible only under a microscope.
  • Full Set of Blood Components on Individual Bases
  • Detailed Guide Describes Each Cell Model and Its Function in the Circulatory System
  • Designed By Wards
This set provides students with a detailed view of blood elements without a microscope. Cast in durable, lightweight plastic and hand painted under the direction of our staff anatomist for realistic features, the eight individual models consist of the erythrocyte, neutrophil, eosinophil, basophil, small lymphocyte, large lymphocyte, monocyte, and platelets.
The models are enlarged 10,000x and are rendered in a centimeter scale that equals micron dimensions of cells in blood smears.
Ordering information: The eight models are mounted on individual stands, and the set includes a teacher’s manual describing each model.