Somso® Chromosome Model
Supplier: MARCUS SOMMER
ZOS 57/4
470029-434EA 779.15 USD
470029-434
Somso® Chromosome Model
Models Molecular Biology Models
Display A Metaphase Chromosome At 10,000X Magnification
  • Shows The Complex Structure Of Chromosomes
  • Removable From Base For Closer Examination
  • 5 Year Manufacturers Warranty
This unique model shows the kinetochores oriented to the outside of the chromosome, as well as the slightly irregular surface of the chromatids. Also shown are the chromatin loops coiled on the inside of the chromosome and the supported or imposed proteins. The lower section of the chromatid is exposed to reveal the chromatin.