Ward's® Terrain Feature Model
Supplier: Plas-Labs
151-TM
470029-382EA 184.8 USD
470029-382
Ward's® Terrain Feature Model
Models Stream Models
Expand the Number of Activities You Can Perform with Your Stream Table
Add the plastic mountain region to the lake area of your stream table to study the movement of water and sediment in relation to a landform; flood the lake region at regular depth increments, then develop a contour map of your observations and measurements. Size: 15"L x 11 1/2"W x 3"H.