Bravais Space Lattice Model Set
Supplier: Klinger
KLINGER EDUCATION PROD CORP SE
470029-192KT 674.67 USD
470029-192
Bravais Space Lattice Model Set
Models Crystal Models
Study the Atomic Arrangements That Make up Most Minerals
  • 14 Models
  • Sturdy Wooden Models
This large, sturdy set of 14 lattice-type models is crafted of wood spheres connected by metal rods, so it won’t break easily, even with frequent handling. Six vibrant colors identify each model with one of the six fundamental crystal systems.
Models:
Triclinic; monoclinic/base-centered; orthorhombic/base, face, body-centered; tetragonal/body-centered; rhombohedral; and hexagonal; cubic/face, body-centered.
Each model is clearly labeled. Size: up to 6".