Fruit Types Riker Mount
Supplier: WHITE OWL ENTERPRISES
R121
470028-132EA 154.95 USD
470028-132
Fruit Types Riker Mount
Specimens Preserved Specimens Display Specimens
Generate Insight into Plant Reproductive Strategies
Excite student interest and generate thought-provoking insight into plant reproductive strategies and seed dispersal with the real specimens attractively displayed in this large Riker mount. Nineteen different specimens are labeled to illustrate the following fruit types; nut, samara, legume, loment, silicle, drupe, berry, capsule, caryopsis, achene, and aggregate. Size: 14" x 21".