Stem Adaptation Riker Mount
Supplier: WHITE OWL ENTERPRISES
R115
470028-130EA 67.95 USD
470028-130
Stem Adaptation Riker Mount
Specimens Preserved Specimens Display Specimens
Learn Plant Morphology by Studying Stem Morphology
Understand how some plant species gain protection, and enhance their reproduction, through modifications of stem structure by using the specimens presented in this 8" x 12" Riker mount. A bulb, corm, and rhizome show stem modifications for reproduction, while stems with thorns, spines, tendrils, and “wings” show adaptations for protection and growth. All specimens are labeled.