Opossum Skull
Supplier: Avantor
470028-022EA 248.95 USD
470028-022
Opossum Skull
Models Zoology Models
This unmounted skull of the common North American Didelphis virginiana shows the extra teeth, which is a characteristic of marsupials.
  • Natural bone
  • Professionally cleaned and prepared
Ordering information: This is a real specimen, so size and appearance may vary from the image.