Ward's® Mantle Xenolith
Supplier: Avantor
470025-896EA 42.59 USD
470025-896
Ward's® Mantle Xenolith
Specimens Rock Specimens Specialty Rocks
Glossy green dunite (olivine) inclusion in non-genetically related basalt. Average size: 3" x 4".