Ward's® Microcline (Pink)
Supplier: Avantor
470025-686PK 9.99 USD
470025-686 470025-688 470025-690
Ward's® Microcline (Pink)
Specimens Mineral Specimens Classroom Minerals
Feldspar group; pink cleavages showing perthitic texture, common potassium feldspar, Mohs’ hardness of 6.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Student specimens, hand specimens, and bulk packs are available.