Geo-logic Sedimentary Rock Set
Supplier: Avantor
470175-720EA 249.99 USD
470175-720 470025-350
Geo-logic Sedimentary Rock Set
Specimens Rock Specimens Sedimentary Rocks
Indicative of Various Sedimentary Formation Processes
  • Developed jointly by experienced educators and Ward's® staff geologists
  • Classroom tested for dependability
  • Designed to save valuables planning and preparation time
When students study the well-defined samples of sedimentary rocks in our GEO-logic set, they will begin to understand the processes by which sedimentary rocks were formed. A teacher’s guide and student copymasters are included for the following activities: categorizing sedimentary rocks by origin, settling and layering, and sediment transport using a stream table. The set is housed in a compartmented collection box.
GEO-logic is a unique, comprehensive series of teaching collections designed to assist both teacher and student in understanding basic earth science concepts through hands-on activities and carefully prepared teacher and student guides.
Contents:
12 individually numbered samples, 1×2"
Magnifier
Dilute HCl (bottle)
Rocks:
breccia, chert, coal (bituminous), conglomerate, coquina, dolostone, gypsum, limestone, sandstone, shale, siltstone, travertine
Caution: Due to the nature of this product it may contain asbestos.