Geo-logic Igneous Rock Set
Supplier: Avantor
470025-348EA 58.09 USD
470025-348
Geo-logic Igneous Rock Set
Specimens Rock Specimens Igneous Rocks
Study the Formation of Different Igneous Rocks
  • Developed jointly by experienced educators and Ward's® staff geologists
  • Classroom tested for dependability
  • Designed to save valuable planning and preparation time
The GEO-logic activities in the set will help students learn the different types of igneous rocks, compare and contrast characteristics, and understand their place in the rock cycle. A teacher's guide and student copymasters are included for the following activities: relating igneous textures to origin, classifying igneous rocks by texture and composition, and estimating percentages of minerals. The set is housed in a compartmented collection box.
GEO-logic is a unique, comprehensive series of teaching collections designed to assist both teacher and student in understanding basic earth science concepts through hands-on activities and carefully prepared teacher and student guides
Contents:
12 individually numbered samples, 1 x 2"
Rocks: anorthosite, basalt, diorite, gabbro, granite, obsidian, peridotite, pumice, rhyolite (2), scoria, syenite
Magnifier