Ward's® AGI/NAGT Laboratory Mineral Set
Supplier: Avantor
470025-290EA 91.79 USD
470025-290
Ward's® AGI/NAGT Laboratory Mineral Set
Specimens Mineral Specimens Mineral Collections
Specimens To Supplement the Most Widely-Used College Lab Manual for Physical Geology
  • Designed to work with Laboratory Manual in Physical Geology (Busch)
  • Hand cut by WARD's Geologists
Improve student understanding of mineral properties and uses with this comprehensive set of important rock-forming and ore minerals customized to complement the testing activities found in your AGI/NAGT Laboratory Manual in Physical Geology. These high-quality samples display excellent consistency and uniformity. Housed in a compartmented collection box, the set also comes with an identification list.
Contents:
24 numbered samples, 1×1" to 1×2"
Ordering information: Mineral contents include albite, augite, bauxite, biotite, calcite, chalcopyrite, dolomite, fluorite, galena, garnet, graphite, gypsum, halite, hematite, hornblende, kaolinite, limonite, magnetite, microcline, quartz, muscovite, olivine, sphalerite, and talc.
Caution: Due to the nature of this product it may contain asbestos.