Ward's® Luster Collection
Supplier: Avantor
470025-260EA 34.79 USD
470025-260
Ward's® Luster Collection
Specimens Mineral Specimens Mineral Collections
Examples of Different Types of Surface Appearance
The minerals in this collection are each perfect examples of one type of luster. The set is housed in a compartmented collection box, and an identification list is included.Contents: 12 numbered samples, 1×2" Minerals: Metallic Luster: pyrite Adamantine Luster: sphalerite Vitreous Luster: quartz Resinous Luster: sulfur Greasy Luster: nepheline Pearly Luster: talcSilky Luster: satin spar Splendent Luster: micaceous hematite Shining Luster: calcite Glistening Luster: prochlorite Submetallic Luster: ilmenite Dull Luster: chalk
Caution: Due to the nature of this product it may contain asbestos.