Minerals of the Earth’s Crust Collection
Supplier: Avantor
470025-188EA 46.49 USD
470025-188
Minerals of the Earth’s Crust Collection
Specimens Mineral Specimens Mineral Collections
The Most Abundant Minerals in the Earth’s Crust
Students can view what’s under their feet simply by looking into one box. A detailed list of the elements and minerals that make up the Earth’s crust is also included, in weight percentages. The set is housed in a compartmented collection box and comes with an identification list.Contents:16 numbered samples, 1×1" Minerals:actinolite, albite, anorthite, augite, biotite, calcite, chlorite, diopside, dolomite, hornblende, kaolin, magnetite, microcline, muscovite, olivine, quartz
Caution: Due to the nature of this product it may contain asbestos.