Ward's® Ultramafic Rocks Thin Sections Set
Supplier: Avantor
470024-960KT 849.29 USD
470024-960
Ward's® Ultramafic Rocks Thin Sections Set
Specimens Rock Specimens Rock Thin Sections
Enhance Your Ultramafic Studies with This Special Petrographic Resource
  • Hand Made by Wards® staff Geologists
  • 12 Specially Selected Samples
  • Microscopic view of Igneous Samples with Very Low Silica Content
Due to the growing demand and interest in ultramafic rocks, we’ve prepared a set of twelve thin sections to match our Ultramafic Rock Collection. Each slide has been selected to illustrate the key textural and compositional features of ultramafic rocks.
Contents: bronzitite, carbonatite, dunite (2), kimberlite, lamproite, peridotite (3), pyroxenite, serpentinite, soapstone