Ward's® Refractive Index Immersion Slide Set
Supplier: Avantor
470024-872KT 222.49 USD
470024-872
Ward's® Refractive Index Immersion Slide Set
Specimens Mineral Specimens Mineral Thin Sections
Ten New Slides in a Complete Teaching Set
  • Hand Made by Wards® staff Geologists
  • 10 Specially Selected Samples
  • Demonstrates Refractive Index of Minerals
Students learn to evaluate refractive index using the Becke Line method.
Set includes teacher’s guide and student worksheets.
10 slides with one mineral each: fluorite, microcline, albite, quartz, labradorite, biotite, calcite, hornblende, almandine, granite.