Mica Set
Supplier: Avantor
470024-862KT 758.39 USD
470024-862
Mica Set
Specimens Mineral Specimens Mineral Collections
Specially selected thin sections of important rock forming minerals. Set may contain more than one example of the same mineral to depict variations in occurrence and distinct mineral associations.
Slides (12); annite, biotite, clinochlore, fuschite, glauconite, lepidolite, margarite, muscovite, paragonite, phlogopite, sericite, zinnwaldite.