Preserved Specimen Conditioning Fluid
Supplier: Avantor
470024-842EA
21.25
USD
470024-842
Preserved Specimen Conditioning Fluid
Chemicals for Science Education
Hazard Info: Toxic
Shelf Life (months): 36
Storage: Blue
This solution can be used as a mold inhibitor for dissection specimens if you choose to dissect your specimen over multiple days/weeks.
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.