Loading Dye
Supplier: ALDON CORP SE
LL0248-5ML
470024-814EA 9.5 USD
470024-814
Loading Dye
Stains Electrophoresis Stains
Solution used for tracking DNA during agarose electrophoresis; prepared with ultra-pure water and reagent-grade chemicals. 10X concentration.