Magnetic Fingerprinting Supplies
Supplier: THE LYNN PEAVEY COMPANY
55575 1003941 55574 1-0175 55571 55577 30009
470002-524EA 17.7 USD
470002-524 470020-206 470024-630 470024-632 470024-634 470024-636 470201-172 470358-934
Magnetic Fingerprinting Supplies
Educational Materials Forensics Educational Materials Forensics Supplies
Apply powder and dust away excess to highlight fingerprints
  • Available in multiple colors
  • Available in two different sizes
Depress the plunger to pick up the magnetic fingerprint powder, dust the surface containing the prints, and then pull up on the plunger again to expel the excess powder into its container.
The larger base of the deluxe applicator allows you to pick up five times more powder than the standard applicator. Length of each: 5¹/₂".
Dual-Use Magnetic Fingerprint Powder: Highlights prints on both dark and light surfaces. Black Magnetic Fingerprint Powder: For highlighting prints on most standard surfaces. Red Magnetic Fingerprint Powder: Fluoresces under UV light. High lights prints on both dark and light surfaces. Deluxe applicator available to U.S. customers only; similar product will ship elsewhere.