Liquid Cyanoacrylate Glue
Supplier: FORENSICS SOURCE
LL-0002
470024-626EA 21.95 USD
470024-626
Liquid Cyanoacrylate Glue
Adhesives
This glue is suitable for use in forensics.
  • Produces hard, white visible impressions
  • 1 oz.
When heated in a closed chamber, the fumes from this “Super Glue” adhesive react with the residues in latent prints to produce hard, white visible impressions.