Ward's® This Old Bone Lab Activity
Supplier: Avantor
470024-250EA 104.09 USD
470024-250
Ward's® This Old Bone Lab Activity
Educational Classroom Kits and Activities
With This Old Bone Lab Activity, students perform an actual technique used by forensic anthropologists to determine the age of individuals at their time of death.
  • For Grades 10-12
  • Students Determine Age from Osteon Counts
  • Estimated Class Time Required: 45 Minutes
  • Teacher's Guide Included
They gain valuable microscopy and anatomy skills in this activity as they learn to count, measure, and analyze the Haversian systems in bone using a compound microscope with an ocular micrometer (both sold separately).
The lab requires 45 minutes.
Ordering information: This kit includes a stage micrometer, six prepared microscope slides of ground human bone, a teacher’s guide, and student copy masters. An ocular micrometer (reticle), a compound microscope, and adhesive tape are required but not included.