Ward's® Phenolphthalein Blood Test Kit
Supplier: Ward's Science
DD6001-KIT
470024-156EA 33.99 USD
470024-156
Ward's® Phenolphthalein Blood Test Kit
Educational Classroom Kits and Activities
Includes a known blood standard to monitor results.
  • Grades 9-12
  • Estimated class/lab time required is 30 minutes
Save time in laboratory experiments using the premixed chemicals and plastic dropper bottles of liquid reagents in this kit. Everything you need for the presumptive determination of real animal blood is included in a plastic storage case, with enough materials to conduct 30 tests. Note: The phenolphthalein test will not distinguish between human and animal blood. Further serology analysis is necessary.