Ward's® Quantifying Enzyme Activity with Amylase Lab Activity
Supplier: Avantor
470024-116KT 79.99 USD
470024-116
Ward's® Quantifying Enzyme Activity with Amylase Lab Activity
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Unique Method Allows Easy Observation of Starch Breakdown
  • Vividly demonstrates enzyme catalysis
  • Materials for 15 lab groups
  • Time Required: ~60 minutes
In this engaging activity, students will vary the concentration of amylase, which is an enzyme that breaks down starch in order to quantify its effects. A vivid, quantifiable result is provided via the use of a starch indicator. It includes enough materials for 15 setups, a teacher’s guide, and student study guide copy-master.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.