The Paper Trail Activity
Supplier: Kemtec
10-544
470024-064EA 296.1 USD
470024-064
The Paper Trail Activity
Educational Materials Forensics Educational Materials Crime Scene Evidence Learning Activities
Use forensics for forgery detection.
  • Grades 5-8
  • Physical and chemical analyses
“Follow the money” using forgery identifying techniques as demonstrated in this activity. Students learn both physical and chemical document analysis by investigating with learned writing techniques, chromatography, and iodine solution. Kit includes all required materials, teacher’s manual, and student guide.