Psychrometer Kit Set/15°C
Supplier: The Science Source
16221
470023-946KT 113 USD
470023-946
Psychrometer Kit Set/15°C
Hygrometers Psychrometers
Measure humidity and temperature with this psychrometer kit.
  • Includes thermometer and hygrometer
  • Ideal for environmental and weather studies
  • –40 to +50 °C
  • 6½×¾" size
Explore Weather Science with the Psychrometer Kit Perfect for hands-on learning, this kit enables students to measure both temperature and humidity—key elements in understanding weather and atmospheric conditions. Every kit features two vinyl-backed alcohol thermometers mounted on durable plastic handles, along with a wick, rubber band, and a vapor pressure/humidity chart for accurate readings.
Delivery information: Kit includes materials to make 15 the psychrometers: 2 thermometers, a handle, wet bulb wick, and psychrometric chart.