Ward's® Protein Fixative Stain
Supplier: ALDON CORP SE
WW0080-500
470023-908EA 29.05 USD
470023-908
Ward's® Protein Fixative Stain
Chemicals for Science Education
Highlights Protein Gels Safely, in 30 Minutes
Our stain images proteins electrophoresed on polyacrylamide gels and, because it’s vinegar based, it’s completely nontoxic. It will destain in water, 10% acetic acid, or white vinegar, eliminating the use of hazardous solutions. The 500 mL bottle stains up to 20 gels.