Molarity Lab Activity
Supplier: Kemtec
13-136
470023-900KT 295.7 USD
470023-900
Molarity Lab Activity
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Use the Mole Concept to Calculate Various Concentrations of Chemical Solutions.
  • Only requires basic labware
  • Determine unknown concentrations using titrations
Determine the concentrations of four solutions of hydrochloric acid in unknown concentrations by titrating them with known concentrations of sodium hydroxide. The kit contains two laboratory exercises, a reproducible manual and enough materials for four setups.