Families of Elements Lab Activity
Supplier: Kemtec
13-132
470023-898KT 561.85 USD
470023-898
Families of Elements Lab Activity
Educational Materials Chemistry Educational Materials Periodic Table Learning Activities
Explore the Similarities Among Groups of Elements
  • Ideal for grades 9-12
  • Includes two experiments
Students are introduced to families (groups) within the periodic table that have the same number of electrons in their outer energy orbitals. Tests are conducted to determine if those elements have similar chemical activity. This kit includes two experiments, one in which students determine the reactivity of the nonmetals in the halogen family and one in which they observe the properties of oxygen. Worksheets focus on identifying characteristics of elements within the halogen, oxygen and nitrogen families, separation methods, and the observed chemical reactions.
Materials Included:
Sodium bromide
Nitric acid
Mineral oil
Potassium iodide
Potassium bromide
Chlorine water
Iodine water
Sodium chloride
Hydrogen peroxide
Manganese dioxide
Glass Vials w/ Caps
Steel Wool
Wood Splints
Materials Needed But Not Included: chemical ventilation hood, general lab supplies. Includes nitric acid.