Kemtec® Tallquist Hemoglobin Scale
Supplier: Kemtec
12-136
470023-872EA 40.45 USD
470023-872
Kemtec® Tallquist Hemoglobin Scale
Test Paper Indicator Paper
Standard hemoglobin reference scale.
  • A fast test to determine anemia levels
  • Paper for 150 tests
The Tallquist hemoglobin scale allows a quick assessment of a patient's hemoglobin levels. Allow blood to absorb into one of the test papers and compare the blood to the color scale to determine the concentration of hemoglobin in blood under normal and anemic conditions.
Delivery information: See datasheet for contents information.