Ward's® World Beneath Our Feet Lab Activity
Supplier: Avantor
470023-828KT 189.09 USD
470023-828
Ward's® World Beneath Our Feet Lab Activity
Educational Classroom Kits and Activities
Look for Macro- and Microorganisms in Soil Samples Your Class Collects
  • Observe soil microorganisms
  • Practice sterile lab techniques
  • Includes a teacher's guide
Under our feet is a whole world of organisms your students might never see. You can show your students this hidden world with our lab activity. After sifting through soil for macroscopic creatures, students use Gram staining techniques and examine the soil for microscopic creatures. The kit comes with enough materials for ten setups, a teacher’s guide, and student copymaster.