Ward's® Halides: Family Characteristics Lab Activity
Supplier: Avantor
470023-824KT 286.59 USD
470023-824
Ward's® Halides: Family Characteristics Lab Activity
Educational Classroom Kits and Activities
Observe the Chemical Properties of Element Families
  • Students explore the periodic table
  • Can be completed in 40-50 minutes
Through the process of combining oxygen and halide family compounds with metal nitrates and copper sulfate, students will observe and compare the chemical reactivities of various elements.
Ordering information: Kit includes Copper Sulfate, Iron (III) Nitrate, Lead Nitrate, Silver Nitrate, Sodium Bromide, Sodium Chloride, Sodium Iodine, Microcentrifuge Tubes with Caps, Microcentrifuge Trays, Pipets, and Acid/Neutral/Base Paper Strips. Masking Tape, PPE are needed but not included.