Ward's® Distinguishing Characteristics of Acids and Bases Lab Activity
Supplier: Avantor
470023-816KT 424.49 USD
470023-816
Ward's® Distinguishing Characteristics of Acids and Bases Lab Activity
Educational Classroom Kits and Activities
Explore the Distinctly Different Properties of Acids and Bases
  • Introduce students to properties of acids and bases
  • Students identify solutions using empirical data
Students will test the included solutions for pH, conductivity, reactivity, and more. They then use this data to identify the chemical characteristics common to acidic, neutral, and alkaline solutions.
Ordering information: Kit includes Copper Sulfate, Hydrochloric Acid, Sodium Hydroxide, Vegetable Oil, Universal Indicator, Magnesium Ribbon, Zinc Strips, 1K ohm Resistors, Battery Connectors, Electrical Tape, Microcentrifuge Tubes, Microcentrifuge Trays, and Stem Pipets. (5) 9-V Batteries are needed but not included.