Ward's® Structure and Function of Mitochondria Lab Activity
Supplier: Avantor
470023-810KT 75.19 USD
470023-810
Ward's® Structure and Function of Mitochondria Lab Activity
Educational Classroom Kits and Activities
Observe the “Energy Producers” in Cells
Observe mitochondria both as cytoplasmic organelles and as centers of enzyme activity. Students will prepare microscope slides using celery and a special stain that acts as a hydrogen acceptor; as the dye loses color, students can actually watch the mitochondria under the microscope as they oxidize food. The kit includes two prepared reference slides of plant and animal mitochondria, enough materials for six setups, a teacher’s guide, and student copymaster.