Ward's® Exploring Stellar Light and Temperature Lab Activity
Supplier: Avantor
470023-714KT 414.39 USD
470023-714
Ward's® Exploring Stellar Light and Temperature Lab Activity
Educational Classroom Kits and Activities
Analyze Star’s Spectra and Calculate Temperatures
Students learn to use a diffraction grating with a variable resistance light board to observe how light is separated into various colors, then determine how a star’s color indicates its temperature. It comes with a teacher’s guide and student copymasters.